missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Disks large homolog 3 Protein

Product Code. 16000221
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Disks large homolog 3, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_113827.1
Gene ID (Entrez) 58948
Name Disks large homolog 3 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Dlg3
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Rat
Species E. coli
Protein Tag GST
Sequence MHKHQHCCKC PECYEVTRLA ALRRLEPPGY GDWQVPDPYG PSGGNGASSG YGGYSSQTLP SQAGATPTPR TKAKLIPTGR DVGPVPPKPV PGKNTPKLNG SGPSWWPECT CTNRDWYEQA SPAPLLVNPE ALEPSLSVNG SDGMFKYEEI VLERGNSGLG FSIAGGIDNP HVPDDPGIFI TKIIPGGAAA MDGRLGVNDC VLRVNEVDVS EVVHSRAVEA LKEAGPVVRL VVRRRQPPPE TIMEVNLLKG PKGLGFSIAG GIGNQHIPGD NSIYITKIIE GGAAQKDGRL QIGDRLLAVN NTNLQDVRHE EAVASLKNTS DMVYLKVAKP GSLHLNDMYA PPDYASTFTA LADNHISHNS SLGYLGAVES KVTYPAPPQV PPTRYSPIPR HMLAEEDFTR EPRKIILHKG STGLGFNIVG GEDGEGIFVS FILAGGPADL SGELRRGDRI LSVNGVNLRN ATHEQAAAAL KRAGQSVTIV AQYRPEEYSR FESKIHDLRE QMMNSSMSSG SGSLRTSEKR SLYVRALFDY DRTRDSCLPS QGLSFSYGDI LHVINASDDE WWQARLVTPH GESEQIGVIP SKKRVEKKER ARLKTVKFHA RTGMIESNRD FPGLSDDYYG AKNLKGVTSN TSDSESSSKG QEDAILSYEP VTRQEIHYAR PVIILGPMKD RVNDDLISEF PHKFGSCVPH TTRPRRDNEV DGQDYHFVVS REQMEKDIQD NKFIEAGQFN DNLYGTSIQS VRAVAERGKH CILDVSGNAI KRLQQAQLYP IAIFIKPKSI EALMEMNRRQ TYEQANKIFD KAMKLEQEFG EYFTAIVQGD SLEEIYNKIK QIIEDQSGHY IWVPSPEKL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Disks large homolog 3 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.