missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Calcium-dependent phospholipase A2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Calcium-dependent phospholipase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6500-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_058870.1 | |
Calcium-dependent phospholipase A2 Protein | |
Research Use Only | |
PLA2G5 | |
Recombinant Protein | |
E. coli | |
GLLELKSMIE KVTGKNAVKN YGFYGCYCGW GGHGTPKDGT DWCCRMHDRC YGLLEEKHCA IRTQSYDYRF TQDLVICEHD SFCPVRLCAC DRKLVYCLRR NLWSYNRLYQ YYPNFLC | |
Greater than 90% as determined by SDS-PAGE. |
29354 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |