missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9349-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088141

  • 5100.00 DKK / 100µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036767.1
Anionic trypsin-1 Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
24691
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Prss1
Recombinant Protein
Yeast
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers