missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Anionic trypsin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9349-ye-1mg

 View more versions of this product

Product Code. 16008151

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_036767.1
Anionic trypsin-1 Protein
Research Use Only
Prss1
Recombinant Protein
Yeast
IVGGYTCPEH SVPYQVSLNS GYHFCGGSLI NDQWVVSAAH CYKSRIQVRL GEHNINVLEG DEQFINAAKI IKHPNYSSWT LNNDIMLIKL SSPVKLNARV APVALPSACA PAGTQCLISG WGNTLSNGVN NPDLLQCVDA PVLSQADCEA AYPGEITSSM ICVGFLEGGK DSCQGDSGGP VVCNGQLQGI VSWGYGCALP
Greater than 90% as determined by SDS-PAGE.
24691
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Anionic trypsin-1 Protein > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.