missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV PhosphoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Phosphoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6952-100ug

 View more versions of this product

Product Code. 16066850

  • 7745.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

RABV PhosphoProtein
Research Use Only
P
Recombinant Protein
E. coli
MSKIFVNPSA IRAGLADLEM AEETVDLINR NIEDNQAHLQ GEPIEVDNLP EDMGRLHLDD GKSPNPGEMA KVGEGKYRED FQMDEGEDPS FLFQSYLENV GVQIVRQMRS GERFLKIWSQ TVEEIISYVA VNFPNPPGKS SEDKSTQTTG RELKKETTPT PSQRESQSSK ARMAAQTASG PPALEWSATN EKDDLSVEAE IAHQIAESFS KKYKFPSRSS GILLYNFEQL KMNLDDIVKE AKNVPGVTRL AHDGSKLPLR CVLGWVALAN SKKFQLLVES DKLSKIMQDD LNRYTSC
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
RABV
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant RABV PhosphoProtein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.