missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV NucleoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7060-ec-200ug
This item is not returnable.
View return policy
Specifications
RABV NucleoProtein | |
Research Use Only | |
N | |
Recombinant Protein | |
E. coli | |
MDADKIVFKV NNQVVSLKPE IIVDQYEYKY PAIKDLKKPC ITLGKAPDLN KAYKSVLSGM NAAKLDPDDV CSYLAAAMQF FEGTCPEDWT SYGILIARKG DRITPNSLVE IKRTDVEGNW ALTGGMELTR DPTVSEHASL VGLLLSLYRL SKISGQNTGN YKTNIADRIE QIFETAPFVK IVEHHTLMTT HKMCANWSTI PNFRFLAGTY DMFFSRIEHL YSAIRVGTVV TAYEDCSGLV SFTGFIKQIN LTAREAILYF FHKNFEEEIR RMFEPGQETA VPHSYFIHFR SLGLSGKSPY SSNAVGHVFN LIHFVGCYMG QVRSLNATVI AACAPHEMSV LGGYLGEEFF GKGTFERRFF RDEKELQEYE AAELTKTDVA LADDGTVNSD DEDYFSGETR SPEAVYTRIM MNGGRLKRSH IRRYVSVSSN HQARPNSFAE FLNKTYSNDS | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
RABV | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant RABV NucleoProtein > 200μg
Spot an opportunity for improvement?Share a Content Correction