missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV GlycoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7425-ye-10ug
This item is not returnable.
View return policy
Specifications
RABV GlycoProtein | |
Research Use Only | |
G | |
Recombinant Protein | |
Yeast | |
KFPIYTIPDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ISAIKVNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYH WLRTVKTTKE SLVIISPSVA DLDPYDKSLH SRVFPSGKCS GITISSTYCS TNHDYTIWMP ENPRLGTSCD IFTNSRGKRA SKGGKTCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSDETK WCPPDQLVNL HDFRSDEIEH LVVEELVKKR EECLDALESI MATKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEIIP SKGCLRVGGR CHPHVNGVFF NGIILGPDGH VLIPEMQSSL LQQHMELLES SVIPLMHPLA DPSTVFKDGD EAEDFVEVHL PDVHKQISGV DLGLPSWGKY | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
RABV | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant RABV GlycoProtein > 10μg
Spot an opportunity for improvement?Share a Content Correction