missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant RABV GlycoProtein

A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7425-ye-50ug

 View more versions of this product

Product Code. 16038820

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

RABV GlycoProtein
Research Use Only
G
Recombinant Protein
Yeast
KFPIYTIPDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ISAIKVNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYH WLRTVKTTKE SLVIISPSVA DLDPYDKSLH SRVFPSGKCS GITISSTYCS TNHDYTIWMP ENPRLGTSCD IFTNSRGKRA SKGGKTCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSDETK WCPPDQLVNL HDFRSDEIEH LVVEELVKKR EECLDALESI MATKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEIIP SKGCLRVGGR CHPHVNGVFF NGIILGPDGH VLIPEMQSSL LQQHMELLES SVIPLMHPLA DPSTVFKDGD EAEDFVEVHL PDVHKQISGV DLGLPSWGKY
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
RABV
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant RABV GlycoProtein > 50μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.