missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig E-Selectin / CD62e / SELE Protein
A DNA sequence encoding the Sus scrofa (Pig) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.
2300.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_999433.1 |
---|---|
Gene ID (Entrez) | 397508 |
Name | E-Selectin / CD62e / SELE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034890
|
enQuireBio™
QP6661-ec-10ug |
10 ÎĽg | N/A | ||||||
16074890
|
enQuireBio™
QP6661-ec-50ug |
50 ÎĽg | N/A | ||||||
16024890
|
enQuireBio™
QP6661-ec-100ug |
100 ÎĽg |
7110.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16054890
|
enQuireBio™
QP6661-ec-200ug |
200 ÎĽg | N/A | ||||||
16064890
|
enQuireBio™
QP6661-ec-500ug |
500 ÎĽg | N/A | ||||||
16044890
|
enQuireBio™
QP6661-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_999433.1 | |
E-Selectin / CD62e / SELE Protein | |
Not determined. | |
Extracellular Domain | |
Pig | |
GST | |
Tris-based buffer, 50% glycerol |
397508 | |
Research Use Only | |
SELE | |
Recombinant Protein | |
E. coli | |
WSYSASTETM TFDDASAYCQ QRYTHLVAIQ NHAEIEYLNS TFNYSASYYW IGIRKINGTW TWIGTKKALT PEATNWAPGE PNNKQSNEDC VEIYIKRDKD SGKWNDERCS KKKLALCYTA ACTPTSCSGH GECIETINSS TCQCYPGFRG LQCEQVVECD ALENPVNGVV TCPQSLPWNT TCAFECKEGF ELIGPEHLQC TSSGSWDGKK PTCKAVTCDT VGHPQNGDVS CNHSSIGEFA YKSTCHFTCA EGFGLQGPAQ IECTAQGQWT QQAPVCKAVK CPAVSQPKNG LVKFTHSPTG EFTYKSSCAF SCEEGFELRG SAQLACTSQG QWTQEVPSCQ VVQCSSLEVP REINMSCSGE PVFGAVCTFA CPEGWMLNGS VALTCGATGH WSGMLPTCEA PAESKIP | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Pig E-Selectin / CD62e / SELE Protein