missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig CALR / Calreticulin Protein
A DNA sequence encoding the Sus scrofa (Pig) CALR / Calreticulin, was expressed in the hosts and tags indicated.
2615.00 DKK - 29485.00 DKK
Specifications
Accession Number | NP_001167604.1 |
---|---|
Gene ID (Entrez) | 100381266 |
Name | CALR / Calreticulin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067481
|
enQuireBio™
QP9027-ye-10ug |
10 ÎĽg | N/A | ||||||
16007491
|
enQuireBio™
QP9027-ye-50ug |
50 ÎĽg | N/A | ||||||
16057481
|
enQuireBio™
QP9027-ye-100ug |
100 ÎĽg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16087481
|
enQuireBio™
QP9027-ye-200ug |
200 ÎĽg | N/A | ||||||
16097481
|
enQuireBio™
QP9027-ye-500ug |
500 ÎĽg | N/A | ||||||
16077481
|
enQuireBio™
QP9027-ye-1mg |
1 mg |
29485.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001167604.1 | |
CALR / Calreticulin Protein | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
100381266 | |
Research Use Only | |
Calr | |
Recombinant Protein | |
Yeast | |
EPTIYFKEQF LDGDGWTDRW IESKHKPDFG RFVLSSGKFY GDQEKDKGLQ TSQDARFYAL SARFEPFSNK GQTLVVQFTV KHEQNIDCGG GYVKLFPDGL DQTDMHGDSE YNIMFGPDIC GPGTKKVHVI FNYKGKNVLI NKDIRCKDDE FTHLYTLIVR PDNTYEVKID NSQVESGSLE DDWDFLPPKK IKDPDAVKPE DWDERAKIDD PTDSKPEDWD KPEHIPDPDA KKPEDWDEEM DGEWEPPVIQ NPEYKGEWKP RQIDNPDYKG TWIHPEIDNP EYSPDSNIYA YENFAVLGLD LWQVKSGTIF DNFLITNDEA YAEEFGNETW GVTKAAEKQM KDKQDEEQRL KEEEEEKKRK EEEEVDKEDE EDKDEDEEEE DEKEEEEEED AAAGQAKDEL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Pig CALR / Calreticulin Protein