missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig C5a / Complement 5a Protein
A DNA sequence encoding the Sus scrofa (Pig) C5a / Complement 5a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9020-ye-500ug
This item is not returnable.
View return policy
Specifications
C5a / Complement 5a Protein | |
Research Use Only | |
C5 | |
Recombinant Protein | |
Yeast | |
MLQKKIEEEA AKYKYAMLKK CCYDGAYRND DETCEERAAR IKIGPKCVKA FKDCCYIANQ VRAEQSHKNI QLGR | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Pig C5a / Complement 5a Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction