Få mere at vide
enQuireBio™ Recombinant other Gly m 4.0101 Protein
Artikelnummer. 15999038
Mængde:
1 mg
100 μg
50 μg
Pakningsstørrelse:
100µg
1mg
50µg
Artikelnummer. 15999038
Brand: enQuireBio™ QP11997100ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A cDNA sequence encoding the Gly m 4.0101 was constructed and used to recombinantly synthesize the protein.
Tekniske data
Navn Gly m 4.0101 Protein
Mængde 100 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttype Recombinant Protein
Krydsreaktivitet Other
Arter Baculovirus
Protein tag His
Sekvens MGVFTFEDEI NSPVAPATLY KALVTDADNV IPKALDSFKS VENVEGNGGP GTIKKITFLE DGETKFVLHK IESIDEANLG YSYSVVGGAA LPDTAEKITF DSKLVAGPNG GSAGKLTVKY ETKGDAEPNQ DELKTGKAKA DALFKAIEAY LLAHPDYN
Buffer Gly m 4.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Renhed eller kvalitetsklasse Greater than 80.0% as determined by SDS-PAGE.