missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma 46 kDa surface antigen Protein

A DNA sequence encoding the Mycoplasma hyopneumoniae (strain 232) 46 kDa surface antigen, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7000-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076980

  • 22530.00 DKK / 1mg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_011206345.1
Greater than 86.9% as determined by SDS-PAGE.
Research Use Only
p46
Recombinant Protein
E. coli
CGQTESGSTS DSKPQAETLK HKVSNDSIRI ALTDPDNPRW ISAQKDIISY VDETEAATST ITKNQDAQNN WLTQQANLSP APKGFIIAPE NGSGVGTAVN TIADKGIPIV AYDRLITGSD KYDWYVSFDN EKVGELQGLS LAAGLLGKED GAFDSIDQMN EYLKSHMPQE TISFYTIAGS QDDNNSQYFY NGAMKVLKEL MKNSQNKIID LSPEGENAVY VPGWNYGTAG QRIQSFLTIN KDPAGGNKIK AVGSKPASIF KGFLAPNDGM AEQAITKLKL EGFDTQKIFV TGQDYNDKAK TFIKDGDQNM TIYKPDKVLG KVAVEVLRVL IAKKNKASRS EVENELKAKL PNISFKYDNQ TYKVQGKNIN TILVSPVIVT KANVDNPDA
Mycoplasma 46 kDa surface antigen Protein
1 mg
Not determined.
Full Length
Mycoplasma
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers