Få mere at vide
R&D Systems™ Recombinant Mouse Wnt-9a Protein
Formulering:
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Beskrivelse
The Recombinant Mouse Wnt-9a Protein is derived from CHO. The Recombinant Mouse Wnt-9a Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. |
| Gen-id (Entrez) | 216795 |
| Molekylvægt (g/mol) | 37 kDa |
| Navn | Wnt-9a |
| Mængde | 25 μg |
| Kilde | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-9a protein Tyr30-Gly365 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Renhed eller kvalitetsklasse | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |