missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Thioredoxin domain-containing protein 12 Protein

A DNA sequence encoding the Mus musculus (Mouse) Thioredoxin domain-containing protein 12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8212-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083311

  • 17510.00 DKK / 500µg
Estimated Shipment: 03-08-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_079610.1
Thioredoxin domain-containing protein 12 Protein
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
66073
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TXNDC12
Recombinant Protein
Yeast
RTGLGKGFGD HIHWRTLEDG KKEAAASGLP LMVIIHKSWC GACKALKPKF AESTEISELS HNFVMVNLED EEEPRDEDFS PDGGYIPRIL FLDPSGKVRP EIINESGNPS YKYFYVSAEQ VVQGMKEAQE RLTGDAFREK HFQDEL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers