missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Thioredoxin domain-containing protein 12 Protein

Product Code. 16093311
50 μg, 50µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Thioredoxin domain-containing protein 12, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_079610.1
Gene ID (Entrez) 66073
Name Thioredoxin domain-containing protein 12 Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TXNDC12
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Yeast
Protein Tag His
Sequence RTGLGKGFGD HIHWRTLEDG KKEAAASGLP LMVIIHKSWC GACKALKPKF AESTEISELS HNFVMVNLED EEEPRDEDFS PDGGYIPRIL FLDPSGKVRP EIINESGNPS YKYFYVSAEQ VVQGMKEAQE RLTGDAFREK HFQDEL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Thioredoxin domain-containing protein 12 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.