missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Sex-determining region Y Protein
A DNA sequence encoding the Mus musculus (Mouse) Sex-determining region Y protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6730-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035694.1 | |
Sex-determining region Y Protein | |
Research Use Only | |
Sry | |
Recombinant Protein | |
E. coli | |
MEGHVKRPMN AFMVWSRGER HKLAQQNPSM QNTEISKQLG CRWKSLTEAE KRPFFQEAQR LKILHREKYP NYKYQPHRRA KVSQRSGILQ PAVASTKLYN LLQWDRNPHA ITYRQDWSRA AHLYSKNQQS FYWQPVDIPT GHLQ | |
Greater than 90% as determined by SDS-PAGE. |
21674 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |