missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Serum amyloid A-3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Serum amyloid A-3 protein, was expressed in the hosts and tags indicated.

1820.00 DKK - 24645.00 DKK

Specifications

Accession Number NP_035445.1
Gene ID (Entrez) 20210
Name Serum amyloid A-3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16098420
View Documents
enQuireBio™
QP7359-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16038430
View Documents
enQuireBio™
QP7359-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16088420
View Documents
enQuireBio™
QP7359-ec-100ug
100 ÎĽg
5555.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16018430
View Documents
enQuireBio™
QP7359-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028430
View Documents
enQuireBio™
QP7359-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16008430
View Documents
enQuireBio™
QP7359-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_035445.1
Serum amyloid A-3 Protein
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
20210
Research Use Only
Saa3
Recombinant Protein
E. coli
RWVQFMKEAG QGSRDMWRAY SDMKKANWKN SDKYFHARGN YDAARRGPGG AWAAKVISDA REAVQKFTGH GAEDSRADQF ANEWGRSGKD PNHFRPAGLP KRY
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Serum amyloid A-3 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.