missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SerpinE1 / PAI-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) SerpinE1 / PAI-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6672-ye-1mg
This item is not returnable.
View return policy
Specifications
SerpinE1 / PAI-1 Protein | |
Research Use Only | |
Serpine1 | |
Recombinant Protein | |
Yeast | |
TLPLRESHTA HQATDFGVKV FQQVVQASKD RNVVFSPYGV SSVLAMLQMT TAGKTRRQIQ DAMGFKVNEK GTAHALRQLS KELMGPWNKN EISTADAIFV QRDLELVQGF MPHFFKLFQT MVKQVDFSEV ERARFIINDW VERHTKGMIN DLLAKGAVDE LTRLVLVNAL YFSGQWKTPF LEASTHQRLF HKSDGSTVSV PMMAQSNKFN YTEFTTPDGL EYDVVELPYQ RDTLSMFIAA PFEKDVHLSA LTNILDAELI RQWKGNMTRL PRLLILPKFS LETEVDLRGP LEKLGMPDMF SATLADFTSL SDQEQLSVAQ ALQKVRIEVN ESGTVASSST AFVISARMAP TEMVIDRSFL FVVRHNPTET ILFMGQVMEP | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse SerpinE1 / PAI-1 Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction