Learn More
enQuireBio™ Recombinant Mouse SDF2 Protein
A DNA sequence encoding the Mus musculus (Mouse) SDF2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6658-ec-100ug
 View more versions of this product
Product Code. 16044870
  • 5555.00 DKK / 100µg
Specifications
NP_033169.3
SDF2 Protein
Research Use Only
SDF2
Recombinant Protein
E. coli
SNMAVVTCGS VVKLLNTRHN VRLHSHDVRY GSGSGQQSVT GVTSVDDSNS YWRIRGKTAT VCERGTPIKC GQPIRLTHIN TGRNLHSHHF TSPLSGNQEV SAFGEEGEGD YLDDWTVLCN GPYWVRDGEV RFKHSSTDVL LSVTGEQYGR PISGQKEVHG MAQPSQNNYW KAMEGIFMKP SELLRAEVHH AEL
Greater than 80% as determined by SDS-PAGE.
20316
100 μg
Not determined.
Full Length of Mature Protein
Mouse
His
Tris-based buffer, 50% glycerol
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Mouse SDF2 Protein > 100μg