missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Mouse Pyridoxal phosphate phosphatase Protein

Artikelnummer. 16003630
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16003630

Brand: enQuireBio™ QP6482ec500ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Mus musculus (Mouse) Pyridoxal phosphate phosphatase, was expressed in the hosts and tags indicated.

Tekniske data

Adgangsnummer NP_064667.2
Gen-id (Entrez) 57028
Navn Pyridoxal phosphate phosphatase Protein
Mængde 500 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol Pdxp
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Mouse
Arter E. coli
Protein tag His-SUMO
Sekvens MARCERLRGA ALRDVLGQAQ GVLFDCDGVL WNGERIVPGA PELLQRLARA GKNTLFVSNN SRRARPELAL RFARLGFAGL RAEQLFSSAL CAARLLRQRL SGPPDASGAV FVLGGEGLRA ELRAAGLRLA GDPGEDPRVR AVLVGYDEQF SFSRLTEACA HLRDPDCLLV ATDRDPWHPL SDGSRTPGTG SLAAAVETAS GRQALVVGKP SPYMFQCITE DFSVDPARTL MVGDRLETDI LFGHRCGMTT VLTLTGVSSL EEAQAYLTAG QRDLVPHYYV ESIADLMEGL ED
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.