missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRKDC Protein
A DNA sequence encoding the Mus musculus (Mouse) PRKDC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6529-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_035289.2 | |
PRKDC Protein | |
Research Use Only | |
PRKDC | |
Recombinant Protein | |
E. coli | |
EYPFLVKGGE DLRQDQRIEQ IFEVMNAILS QDAACSQRNM QLRTYRVVPM TSRLGLIEWI ENTMTLKDLL LSNMSQEEKV ANNSDPKAPI RDYKDWLMKV SGKSDAGAYV LMYSRANRTE TVVAFRRRES QVPPDLLKRA FVKMSTSPEA FLALRSHFAS SHALLCISHW LLGIGDRHLN NFMVAMETGS VIGIDFGHAF GSATQFLPVP ELMPFRLTRQ FVSLMLPMKE TGLMCTVMVH ALRAFRSCAG LLTDTMEIFV KEPSFDWKS | |
Greater than 90% as determined by SDS-PAGE. |
19090 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |