missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PRKDC Protein

A DNA sequence encoding the Mus musculus (Mouse) PRKDC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6529-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073950

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_035289.2
PRKDC Protein
Research Use Only
PRKDC
Recombinant Protein
E. coli
EYPFLVKGGE DLRQDQRIEQ IFEVMNAILS QDAACSQRNM QLRTYRVVPM TSRLGLIEWI ENTMTLKDLL LSNMSQEEKV ANNSDPKAPI RDYKDWLMKV SGKSDAGAYV LMYSRANRTE TVVAFRRRES QVPPDLLKRA FVKMSTSPEA FLALRSHFAS SHALLCISHW LLGIGDRHLN NFMVAMETGS VIGIDFGHAF GSATQFLPVP ELMPFRLTRQ FVSLMLPMKE TGLMCTVMVH ALRAFRSCAG LLTDTMEIFV KEPSFDWKS
Greater than 90% as determined by SDS-PAGE.
19090
10 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers