missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Oncostatin M (OSM) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Mouse Oncostatin M (OSM) Protein is derived from E. coli. The Recombinant Mouse Oncostatin M (OSM) Protein has been validated for the following applications: Bioactivity.

Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 18413 |
Molecular Weight (g/mol) | 21 kDa |
Name | Oncostatin M/OSM |
Quantity | 25 μg |
Source | E. coli-derived mouse Oncostatin M/OSM protein Ala24-Arg206,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Mouse Oncostatin M (OSM) Protein >
Spot an opportunity for improvement?Share a Content Correction