Få mere at vide
R&D Systems™ Recombinant Mouse NTB-A/SLAMF6 Fc Chimera Protein
Artikelnummer. 18127210
Mængde:
50 μg
Artikelnummer. 18127210
Brand: R&D Systems™ 3986NT050
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
The Recombinant Mouse NTB-A/SLAMF6 Fc Chimera Protein is derived from NS0. The Recombinant Mouse NTB-A/SLAMF6 Fc Chimera Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY
Tekniske data
Formulering Lyophilized from a 0.2μm filtered solution in PBS.
Molekylvægt (g/mol) 50.0 kDa (monomer)
Navn NTB-A/SLAMF6 Fc Chimera
Mængde 50 μg
Kilde Mouse myeloma cell line,NS0-derived mouse NTB-A/SLAMF6 protein, (N-terminus) Mouse NTB-A (Glu31-Asn239) Accession &Num; Q9ET39 IEGRMD Mouse IgG2a (Glu98-Lys330) (C-terminus)
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Disulfide-linked homodimer
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.