missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nqo2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nqo2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8176-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001156713.1 | |
Nqo2 Protein | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
E. coli | |
MAGKKVLIVY AHQEPKSFNG SLKKVAVEEL SKQGCTVTVS DLYSMNFEPR ATRNDITGAP SNPDVFSYGI ETHEAYKKKA LTSDIFEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCRGFAFDIP GFYDSGFLKG KLALLSLTTG GTAEMYTKDG VSGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGLDVSSEEE RKVMLASWAQ RLKSIWKEEP IHCTPPWYFQ E | |
Greater than 90% as determined by SDS-PAGE. |
18105 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Nqo2 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction