missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nqo2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nqo2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8176-ye-1mg

 View more versions of this product

Product Code. 16053081

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001156713.1
Nqo2 Protein
Research Use Only
Nqo2
Recombinant Protein
Yeast
MAGKKVLIVY AHQEPKSFNG SLKKVAVEEL SKQGCTVTVS DLYSMNFEPR ATRNDITGAP SNPDVFSYGI ETHEAYKKKA LTSDIFEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCRGFAFDIP GFYDSGFLKG KLALLSLTTG GTAEMYTKDG VSGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGLDVSSEEE RKVMLASWAQ RLKSIWKEEP IHCTPPWYFQ E
Greater than 90% as determined by SDS-PAGE.
18105
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Nqo2 Protein > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.