missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NQO1 / DT-diaphorase Protein
A DNA sequence encoding the Mus musculus (Mouse) NQO1 / DT-diaphorase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9801-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032732.3 | |
NQO1 / DT-diaphorase Protein | |
Research Use Only | |
Nqo1 | |
Recombinant Protein | |
Yeast | |
AARRALIVLA HSEKTSFNYA MKEAAVEALK KRGWEVLESD LYAMNFNPII SRNDITGELK DSKNFQYPSE SSLAYKEGRL SPDIVAEHKK LEAADLVIFQ FPLQWFGVPA ILKGWFERVL VAGFAYTYAA MYDNGPFQNK KTLLSITTGG SGSMYSLQGV HGDMNVILWP IQSGILRFCG FQVLEPQLVY SIGHTPPDAR MQILEGWKKR LETVWEETPL YFAPSSLFDL NFQAGFLMKK EVQEEQKKNK FGLSVGHHLG KSIPADNQIK ARK | |
Greater than 90% as determined by SDS-PAGE. |
18104 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |