missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MSLN / Mesothelin Protein

A DNA sequence encoding the Mus musculus (Mouse) MSLN / Mesothelin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6389-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092960

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_061345.1
MSLN / Mesothelin Protein
Research Use Only
Msln
Recombinant Protein
Yeast
DAEQKACPPG KEPYKVDEDL IFYQNWELEA CVDGTMLARQ MDLVNEIPFT YEQLSIFKHK LDKTYPQGYP ESLIQQLGHF FRYVSPEDIH QWNVTSPDTV KTLLKVSKGQ KMNAQAIALV ACYLRGGGQL DEDMVKALGD IPLSYLCDFS PQDLHSVPSS VMWLVGPQDL DKCSQRHLGL LYQKACSAFQ NVSGLEYFEK IKTFLGGASV KDLRALSQHN VSMDIATFKR LQVDSLVGLS VAEVQKLLGP NIVDLKTEED KSPVRDWLFR QHQKDLDRLG LGLQGGIPNG YLVLDFNVRE AFS
Greater than 90% as determined by SDS-PAGE.
56047
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers