missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MSLN / Mesothelin Protein
A DNA sequence encoding the Mus musculus (Mouse) MSLN / Mesothelin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6389-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_061345.1 | |
MSLN / Mesothelin Protein | |
Research Use Only | |
Msln | |
Recombinant Protein | |
Yeast | |
DAEQKACPPG KEPYKVDEDL IFYQNWELEA CVDGTMLARQ MDLVNEIPFT YEQLSIFKHK LDKTYPQGYP ESLIQQLGHF FRYVSPEDIH QWNVTSPDTV KTLLKVSKGQ KMNAQAIALV ACYLRGGGQL DEDMVKALGD IPLSYLCDFS PQDLHSVPSS VMWLVGPQDL DKCSQRHLGL LYQKACSAFQ NVSGLEYFEK IKTFLGGASV KDLRALSQHN VSMDIATFKR LQVDSLVGLS VAEVQKLLGP NIVDLKTEED KSPVRDWLFR QHQKDLDRLG LGLQGGIPNG YLVLDFNVRE AFS | |
Greater than 90% as determined by SDS-PAGE. |
56047 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse MSLN / Mesothelin Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction