missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MSLN / Mesothelin Protein
A DNA sequence encoding the Mus musculus (Mouse) MSLN / Mesothelin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6389-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_061345.1 | |
MSLN / Mesothelin Protein | |
Research Use Only | |
Msln | |
Recombinant Protein | |
Yeast | |
DAEQKACPPG KEPYKVDEDL IFYQNWELEA CVDGTMLARQ MDLVNEIPFT YEQLSIFKHK LDKTYPQGYP ESLIQQLGHF FRYVSPEDIH QWNVTSPDTV KTLLKVSKGQ KMNAQAIALV ACYLRGGGQL DEDMVKALGD IPLSYLCDFS PQDLHSVPSS VMWLVGPQDL DKCSQRHLGL LYQKACSAFQ NVSGLEYFEK IKTFLGGASV KDLRALSQHN VSMDIATFKR LQVDSLVGLS VAEVQKLLGP NIVDLKTEED KSPVRDWLFR QHQKDLDRLG LGLQGGIPNG YLVLDFNVRE AFS | |
Greater than 90% as determined by SDS-PAGE. |
56047 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |