missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Recombinant Mouse Mast Cell Protease-6/Mcpt6 Protein

Artikelnummer. 18108500
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18108500 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18108500 Leverandør R&D Systems™ Leverandørnr. 3736SE010

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >500 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P21845
Til brug med (applikation) Enzyme Activity
Formulering Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gen-id (Entrez) 17229
Molekylvægt (g/mol) M.W. (Observed): 37 kDa, reducing conditions; M.W. (theoretical): 30 kDa
Navn Mast Cell Protease-6/Mcpt6
Mængde 10 μg
Kilde Mouse myeloma cell line, NS0-derived mouse Mast Cell Protease-6/Mcpt6 protein Ala22-Ser276, with a C-terminal 10-His tag
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Strukturel form Pro form
Endotoksin koncentration <1.0 EU / 1 μg of the protein by the LAL method.
Gen symbol Tpsb2
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Mast Cell Protease-6/Mcpt6
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.