missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Recombinant Mouse Mast Cell Protease-1/Mcpt1 Protein

Artikelnummer. 18106560
Skift visning
Klik for at se tilgængelige muligheder
:
10ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
18106560 10ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18106560 Leverandør R&D Systems™ Leverandørnr. 5146SE010

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Mast Cell Protease-1/Mcpt1 Protein is derived from NS0. The Recombinant Mouse Mast Cell Protease-1/Mcpt1 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Tekniske data

Formulering Supplied as a 0.2μm filtered solution in Tris and NaCl.
Gen-id (Entrez) 17224
Molekylvægt (g/mol) 27 kDa
Navn Mast Cell Protease-1/Mcpt1
Mængde 10μg
Kilde Mouse myeloma cell line,NS0-derived mouse Mast Cell Protease-1/Mcpt1 protein Glu19-Lys246,with a C-terminal 10-His tag
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Strukturel form Pro form
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.