Learn More
enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7360-ye-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16088440
  • 22530.00 DKK / 1mg
Estimated Shipment: 31-07-2023
to see stock.
Specifications
NP_001157998.1
Major urinary protein 11 Protein
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
100039028
Greater than 81.5% as determined by SDS-PAGE.
Research Use Only
Mup11
Recombinant Protein
Yeast
REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E