missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant Mouse Major urinary protein 11 Protein

Artikelnummer. 16068440
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16068440

missing translation for 'mfr': enQuireBio™ QP7360ye100ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Major urinary protein 11, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer NP_001157998.1
Gen-id (Entrez) 100039028
Navn Major urinary protein 11 Protein
Mængde 100 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol Mup11
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet Mouse
Arter Yeast
Protein tag His
Sekvens REKINGEWHT IILASDKREK IEDNGNFRLF LEQIHVLENS LVLKFHTVRD EECSELSMVA DKTEKAGEYS VTYDGFNTFT IPKTDYDNFL MAHLINEKDG ETFQLMGLYG REPDLSSDIK ERFAQLCEEH GILRENIIDL SNANRCLQAR E
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 81.5% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.