missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lysyl oxidase homolog 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Lysyl oxidase homolog 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7930-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_444313.3 | |
Lysyl oxidase homolog 4 Protein | |
Research Use Only | |
Loxl4 | |
Recombinant Protein | |
Yeast | |
QSSGTKKLRL VGPTDRPEEG RLEVLHQGQW GTVCDDDFAL QEATVACRQL GFESALTWAH SAKYGQGEGP IWLDNVRCLG TEKTLDQCGS NGWGVSDCRH SEDVGVVCHP RRQHGYHSEK VSNALGPQGR RLEEVRLKPI LASAKRHSPV TEGAVEVRYD GHWRQVCDQG WTMNNSRVVC GMLGFPSQTS VNSHYYRKVW NLKMKDPKSR LNSLTKKNSF WIHRVDCLGT EPHLAKCQVQ VAPGRGKLRP ACPGGMHAVV SCVAGPHFRR QKPKPTRKES HAEELKVRLR SGAQVGEGRV EVLMNRQWGT VCDHRWNLIS ASVVCRQLGF GSAREALFGA QLGQGLGPIH LSEVRCRGYE RTLGDCLALE GSQNGCQHAN DAAVRCNIPD MGFQNKVRLA GGRNSEEGVV EVQVEVNGVP RWGTVCSDHW GLTEAMVTCR QLGLGFANFA LKDTWYWQGT PEAKEVVMSG VRCSGTEMAL QQCQRHGPVH CSHGPGRFSA GVACMNSAPD LVMNAQLVQE TAYLEDRPLS MLYCAHEENC LSKSADHMDW PYGYRRLLRF SSQIYNLGRA DFRPKAGRHS WIWHQCHRHY HSIEVFTHYD LLTLNGSKVA EGHKASFCLE DTNCPSGVQR RYACANFGEQ GVAVGCWDTY RHDIDCQWVD ITDVGPGDYI FQVVVNPTND VAESDFSNNM IRCRCKYDGQ RVWLHNCHTG DSYRANAELS LEQEQRLRNN LI | |
Greater than 90% as determined by SDS-PAGE. |
67573 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |