Få mere at vide
R&D Systems™ Recombinant Mouse Lymphotoxin alpha2/beta1 Protein
Artikelnummer. 18107630
Formulering:
Lyophilized from a 0.2μm filtered solution in PBS.
Artikelnummer. 18107630
Brand: R&D Systems™ 1008LY010/CF
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Tekniske data
Formulering Lyophilized from a 0.2μm filtered solution in PBS.
Gen-id (Entrez) 16992
Molekylvægt (g/mol) 18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
Navn Lymphotoxin α2/β1
Mængde 10 μg
Kilde Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Lymphotoxin protein Leu34-Leu202 (LT alpha ) & Gly136-Gly306 (LT beta )
Opbevaringskrav Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Strukturel form Noncovalently-linked heterotrimer
Konjugeret Unconjugated
Rekombinant Recombinant
Renhed eller kvalitetsklasse 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.