missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse KEAP1 / INRF2 Protein
A DNA sequence encoding the Mus musculus (Mouse) KEAP1 / INRF2, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_001103775.1 |
---|---|
Gene ID (Entrez) | 50868 |
Name | KEAP1 / INRF2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16061990
|
enQuireBio™
QP6259-ec-10ug |
10 ÎĽg | N/A | ||||||
16002000
|
enQuireBio™
QP6259-ec-50ug |
50 ÎĽg | N/A | ||||||
16051990
|
enQuireBio™
QP6259-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16081990
|
enQuireBio™
QP6259-ec-200ug |
200 ÎĽg | N/A | ||||||
16091990
|
enQuireBio™
QP6259-ec-500ug |
500 ÎĽg | N/A | ||||||
16071990
|
enQuireBio™
QP6259-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001103775.1 | |
KEAP1 / INRF2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
50868 | |
Research Use Only | |
Keap1 | |
Recombinant Protein | |
E. coli | |
MQPEPKLSGA PRSSQFLPLW SKCPEGAGDA VMYASTECKA EVTPSQDGNR TFSYTLEDHT KQAFGVMNEL RLSQQLCDVT LQVKYEDIPA AQFMAHKVVL ASSSPVFKAM FTNGLREQGM EVVSIEGIHP KVMERLIEFA YTASISVGEK CVLHVMNGAV MYQIDSVVRA CSDFLVQQLD PSNAIGIANF AEQIGCTELH QRAREYIYMH FGEVAKQEEF FNLSHCQLAT LISRDDLNVR CESEVFHACI DWVKYDCPQR RFYVQALLRA VRCHALTPRF LQTQLQKCEI LQADARCKDY LVQIFQELTL HKPTQAVPCR APKVGRLIYT AGGYFRQSLS YLEAYNPSNG SWLRLADLQV PRSGLAGCVV GGLLYAVGGR NNSPDGNTDS SALDCYNPMT NQWSPCASMS VPRNRIGVGV IDGHIYAVGG SHGCIHHSSV ERYEPERDEW HLVAPMLTRR IGVGVAVLNR LLYAVGGFDG TNRLNSAECY YPERNEWRMI TPMNTIRSGA GVCVLHNCIY AAGGYDGQDQ LNSVERYDVE TETWTFVAPM RHHRSALGIT VHQGKIYVLG GYDGHTFLDS VECYDPDSDT WSEVTRMTSG RSGVGVAVTM EPCRKQIDQQ NCTC | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse KEAP1 / INRF2 Protein