missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Kallikrein 7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7688-SE-010
Additional Details : Weight : 0.00970kg
Description
The Recombinant Mouse Kallikrein 7 Protein is derived from NS0. The Recombinant Mouse Kallikrein 7 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in HEPES and NaCl. | |
26 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Recombinant |
23993 | |
Kallikrein 7 | |
Mouse myeloma cell line,NS0-derived mouse Kallikrein 7 protein Gln22-Arg249 with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |