missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein Product Code.: 16047390

enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein

Product Code. 16047390
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16047390

Brand: enQuireBio™ QP7102200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) Kallikrein 1-related peptidase b22, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_034244.1
Gene ID (Entrez) 13646
Name Kallikrein 1-related peptidase b22 Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Klk1b22
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence ILGGFKCEKN SQPWQVAVYY LDEYLCGGVL LDRNWVLTAA HCYEDKYNIW LGKNKLFQDE PSAQHRLVSK SFPHPDFNMS LLQSVPTGAD LSNDLMLLRL SKPADITDVV KPIDLPTTEP KLGSTCLASG WGSINQLIYQ NPNDLQCVSI KLHPNEVCVK AHILKVTDVM LCAGEMNGGK DTCKGDSGGP LICDGVLQGI TSWGSTPCGE PNAPAIYTKL IKFTSWIKDT MAKNP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.