missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IFNAR2 / IFNABR Protein

A DNA sequence encoding the Mus musculus (Mouse) IFNAR2 / IFNABR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9199-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057831

  • 5015.00 DKK / 100µg
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001103968.1
IFNAR2 / IFNABR Protein
Research Use Only
Ifnar2
Recombinant Protein
Yeast
SLETITPSAF DGYPDEPCTI NITIRNSRLI LSWELENKSG PPANYTLWYT VMSKDENLTK VKNCSDTTKS SCDVTDKWLE GMESYVVAIV IVHRGDLTVC RCSDYIVPAN APLEPPEFEI VGFTDHINVT MEFPPVTSKI IQEKMKTTPF VIKEQIGDSV RKKHEPKVNN VTGNFTFVLR DLLPKTNYCV SLYFDDDPAI KSPLKCIVLQ PGQESGLSES A
Greater than 90% as determined by SDS-PAGE.
15976
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers