missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Gp2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Gp2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8169-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_080265.2 | |
Gp2 Protein | |
Research Use Only | |
Gp2 | |
Recombinant Protein | |
E. coli | |
STIHQGYGRP RNSSNLDLDC GSPDSPSSGI CFDPCQNHTV LNDPTRSTEN NDSSVAWCDD NLHGWYRFVG DGGVKMPETC VSVFRCHTSA PMWLSGSHPI LGDGIVSHTA CANWNENCCF WRSEVQVKAC SEELGEYHVY KLQGTPECSL RYCTDPSTAP KNCEITCRPE EECVFQNNNW SCVCRQDLHV SDSQSLQPLL DCGDNEIKVK LDKCLLGGMG FKEEIIAYLN DRNCNGTMQD EPNNWVSMTS PVVANYCGNI LEKNGTHAIY RNTLSLATDF IIRDFRVNVN FQCAYPLDMS VSLETALQPI VSSLTVDVDG AGEFNVKMAL FQDQSYTNPY EGAEVLLPVE SILYVGVLLN RGDTSRFKLL LTNCYATPSE DRHDPVKYFI IKNRCPNQRD STINVRENGV SSESRFSVQM FMFAGNYDLV FLHCEVYLCD STTEQCQPSC STNRLRSSRP AIDYNRVLDL GPITKRSAQS SATSKGTPHT TGFLLAWPMF FLPVFLALLF | |
Greater than 90% as determined by SDS-PAGE. |
67133 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Gp2 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction