missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glypican-4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Glypican-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6110-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070870

  • 14490.00 DKK / 500µg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032176.2
Glypican-4 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
GST
Tris-based buffer, 50% glycerol
14735
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Gpc4
Recombinant Protein
Yeast
AGGAHAEAKP YLLAALCILF LAVQGEWR
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers