missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glypican-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Glypican-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6110-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032176.2 | |
Glypican-4 Protein | |
Research Use Only | |
Gpc4 | |
Recombinant Protein | |
Yeast | |
AGGAHAEAKP YLLAALCILF LAVQGEWR | |
Greater than 90% as determined by SDS-PAGE. |
14735 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |