missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Glucose-6-phosphate isomerase Protein

A DNA sequence encoding the Mus musculus (Mouse) Glucose-6-phosphate isomerase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9161-ye-1mg

 View more versions of this product

Product Code. 16067751

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_032181.2
Glucose-6-phosphate isomerase Protein
Research Use Only
Gpi
Recombinant Protein
Yeast
AALTRNPQFQ KLLEWHRANS ANLKLRELFE ADPERFNNFS LNLNTNHGHI LVDYSKNLVN KEVMQMLVEL AKSRGVEAAR DNMFSGSKIN YTENRAVLHV ALRNRSNTPI KVDGKDVMPE VNRVLDKMKS FCQRVRSGDW KGYTGKSITD IINIGIGGSD LGPLMVTEAL KPYSKGGPRV WFVSNIDGTH IAKTLASLSP ETSLFIIASK TFTTQETITN AETAKEWFLE AAKDPSAVAK HFVALSTNTA KVKEFGIDPQ NMFEFWDWVG GRYSLWSAIG LSIALHVGFD HFEQLLSGAH WMDQHFLKTP LEKNAPVLLA LLGIWYINCY GCETHALLPY DQYMHRFAAY FQQGDMESNG KYITKSGARV DHQTGPIVWG EPGTNGQHAF YQLIHQGTKM IPCDFLIPVQ TQHPIRKGLH HKILLANFLA QTEALMKGKL PEEARKELQA AGKSPEDLEK LLPHKVFEGN RPTNSIVFTK LTPFILGALI AMYEHKIFVQ GIMWDINSFD QWGVELGKQL AKKIEPELEG SSAVTSHDSS TNGLISFIKQ QRDTKLE
Greater than 90% as determined by SDS-PAGE.
14751
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Glucose-6-phosphate isomerase Protein > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.