missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galectin-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Galectin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9839-ye-500ug
This item is not returnable.
View return policy
Specifications
NP_034836.1 | |
Galectin-4 Protein | |
Research Use Only | |
Lgals4 | |
Recombinant Protein | |
Yeast | |
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSVYIQGMAK ENMRRFHVNF AVGQDDGADV AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGKH FELVFMVMPE HYKVVVNGNS FYEYGHRLPV QMVTHLQVDG DLELQSINFL GGQPAAAPYP GAMTIPAYPA GSPGYNPPQM NTLPVMTGPP VFNPRVPYVG ALQGGLTVRR TIIIKGYVLP TARNFVINFK VGSSGDIALH LNPRIGDSVV RNSFMNGSWG AEERKVAYNP FGPGQFFDLS IRCGMDRFKV FANGQHLFDF SHRFQAFQMV DTLEINGDIT LSYVQI | |
Greater than 90% as determined by SDS-PAGE. |
16855 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Galectin-4 Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction