missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein

A DNA sequence encoding the Mus musculus (Mouse) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7503-ye-100ug

 View more versions of this product

Product Code. 16069040

  • 5555.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001295215.1
Fragile histidine triad / FHIT Protein
Research Use Only
Fhit
Recombinant Protein
Yeast
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFRDLH PDEVADLFQV TQRVGTVVEK HFQGTSITFS MQDGPEAGQT VKHVHVHVLP RKAGDFPRND NIYDELQKHD REEEDSPAFW RSEKEMAAEA EALRVYFQA
Greater than 90% as determined by SDS-PAGE.
14198
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.