missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fibroblast growth factor 15 Protein
A DNA sequence encoding the Mus musculus (Mouse) Fibroblast growth factor 15, was expressed in the hosts and tags indicated.
1820.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_032029.1 |
---|---|
Gene ID (Entrez) | 14170 |
Name | Fibroblast growth factor 15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098930
|
enQuireBio™
QP7474-ec-10ug |
10 ÎĽg | N/A | ||||||
16038940
|
enQuireBio™
QP7474-ec-50ug |
50 ÎĽg | N/A | ||||||
16088930
|
enQuireBio™
QP7474-ec-100ug |
100 ÎĽg |
5555.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018940
|
enQuireBio™
QP7474-ec-200ug |
200 ÎĽg | N/A | ||||||
16028940
|
enQuireBio™
QP7474-ec-500ug |
500 ÎĽg | N/A | ||||||
16008940
|
enQuireBio™
QP7474-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_032029.1 | |
Fibroblast growth factor 15 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
14170 | |
Research Use Only | |
Fgf15 | |
Recombinant Protein | |
E. coli | |
RPLAQQSQSV SDEDPLFLYG WGKITRLQYL YSAGPYVSNC FLRIRSDGSV DCEEDQNERN LLEFRAVALK TIAIKDVSSV RYLCMSADGK IYGLIRYSEE DCTFREEMDC LGYNQYRSMK HHLHIIFIQA KPREQLQDQK PSNFIPVFHR SFFETGDQLR SKMFSLPLES DSMDPFRMVE DVDHLVKSPS FQK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Fibroblast growth factor 15 Protein