missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Mouse FCRL1/FcRH1 Protein
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Beskrivelse
The Recombinant Mouse FCRL1/FcRH1 Protein is derived from NS0. The Recombinant Mouse FCRL1/FcRH1 Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-id (Entrez) | 229499 |
| Molekylvægt (g/mol) | 22.5 kDa (monomer) |
| Navn | FCRL1/FcRH1 |
| Mængde | 50 μg |
| Kilde | Mouse myeloma cell line,NS0-derived mouse FCRL1/FcRH1 protein Met1-Ser219,with a C-terminal 6-His tag |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturel form | Disulfide-linked homodimer |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion