missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Mouse EGF Protein
Klik for at se tilgængelige muligheder
Mængde:
200 μg
Beskrivelse
Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED50 for this effect is 20-100 pg/mL.
Tekniske data
Tekniske data
| Adgangsnummer | NP_034243 |
| Til brug med (applikation) | Bioactivity |
| Formulering | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. |
| Gen-id (Entrez) | 1950 |
| Molekylvægt (g/mol) | M.W. (theoretical): 6.2 kDa |
| Navn | EGF |
| Mængde | 200 μg |
| Kilde | E. coli-derived mouse EGF protein Asn977-Arg1029, with an N-terminal Met |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoksin koncentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion